missing translation for 'onlineSavingsMsg'
Learn More

Denatonium benzoate

Catalog No. AAJ6104814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6104814 25 g
AAJ6104803 1 g
AAJ6104806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6104814 Supplier Thermo Scientific Chemicals Supplier No. J6104814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The bitterest compound known | CAS: 3734-33-6 | C28H34N2O3

The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.

Solubility
Soluble in isopropyl alcohol, methanol. Slightly soluble in acetone. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents. Moisture sensitive.

Chemical Identifiers

CAS 3734-33-6
Molecular Formula C28H34N2O3
Synonym unii-m5ba6gaf1o

Specifications

Quantity 25 g
Solubility Soluble in isopropyl alcohol,methanol. Slightly soluble in acetone. Insoluble in ether.
Melting Point 164°C to 169°C
Formula Weight 446.59
Physical Form Solid
Color White
Chemical Name or Material Denatonium benzoate
Hazard Category Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, Causes skin irritation
EINECSNumber 223-095-2
Recommended Storage Ambient

RUO – Research Use Only