accessibility menu, dialog, popup

UserName

H3CNCH3ONHCH3H3COO

CAS RN 3734-33-6

IUPAC Name: benzyl({[(2,6-dimethylphenyl)carbamoyl]methyl})diethylazanium benzoate
Synonyms: denatonium denatonium benzoate 苯甲地那铵
Molecular Weight (g/mol): 446.59
Molecular Formula: C28H34N2O3
InChi Key: VWTINHYPRWEBQY-UHFFFAOYSA-N
SMILES: [O-]C(=O)C1=CC=CC=C1.CC[N+](CC)(CC(=O)NC1=C(C)C=CC=C1C)CC1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)