Learn More
Waters Limited DeltaPak C18 Radial-Pak Column, 100, 15 m, 8 mm X 100 mm, 1/pk
Numéro de catalogue. 50785225
Encompass
Delta-Pak packings, ideal for the separation of peptides, proteins, and natural products, are based on a highly stable, bonded, endcapped 5m or 15m spherical silica. Delta-Pak is available in two different pore sizes (100 and 300) with either C4 or C18 bonded phase. Delta-Pak packings are available in different cartridge and column configurations, providing consistent and predictable scale-up capabilities for milligram-to-gram scale purification. Also, by having the same chemistry available on a high-efficiency 5m particle, you can easily check the purity of your fractions.
Numéro de catalogue. 50785225 Fournisseur Waters Limited Code fournisseur WAT025846
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Delta-Pak packings, ideal for the separation of peptides, proteins, and natural products, are based on a highly stable, bonded, endcapped 5m or 15m spherical silica. Delta-Pak is available in two different pore sizes (100 and 300) with either C4 or C18 bonded phase. Delta-Pak packings are available in different cartridge and column configurations, providing consistent and predictable scale-up capabilities for milligram-to-gram scale purification. Also, by having the same chemistry available on a high-efficiency 5m particle, you can easily check the purity of your fractions.