missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab™ Deionized Water
Description
- Water is purified in-house in the StatLab™ state-of-the-art deionized water purification system
- Incoming water is filtered and softened before it undergoes a reverse osmosis process
- Water is further purified by UV, followed by an ultra-purification ion exchange process
Spécifications
Spécifications
| Color | Clear |
| Grade | Reagent |
| Sterility | Non-sterile |
| Quantity | 1 gal./Bottle, 4 Bottles/Cs. |