missing translation for 'onlineSavingsMsg'
Learn More

Dehydroabietylamine 55.0+%, TCI America™

Catalog No. D1180500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1180500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1180500G Supplier TCI America Supplier No. D1180500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1446-61-3
Molecular Formula C20H32N
Molecular Weight (g/mol) 286.48
MDL Number MFCD00213430
InChI Key JVVXZOOGOGPDRZ-SLFFLAALSA-O
Synonym dehydroabietylamine, +-dehydroabietylamine, 1r,4as,10ar-7-isopropyl-1,4a-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthren-1-yl methanamine, amine d, dehydroabiethylamine, leelamine, podocarpa-8,11,13-trien-15-amine, 13-isopropyl, 1-phenanthrenemethanamine, 1,2,3,4,4a,9,10,10a-octahydro-1,4a-dimethyl-7-1-methylethyl-, 1r,4as,10ar, pubchem7951, 4-12-00-03005 beilstein handbook reference
PubChem CID 62034

Specifications

Boiling Point 344°C
Quantity 500 g
Formula Weight 285.48
Percent Purity ≥55.0% (GC)
Physical Form Liquid
Chemical Name or Material Dehydroabietylamine
EINECSNumber (4)-1242&(7)-0939
TSCA No