missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Defender 6000 Extreme Washdown Scale Indicator

Catalog No. 01919431
Change view
Click to view available options
Construction:
Polycarbonate, Stainless Steel (316)
Stainless Steel (316)
Model:
i-DT61PW AM
i-DT61PW AM Front Installation
i-DT61XWE AM
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Construction Model
01919431 Stainless Steel (316) i-DT61XWE AM
01919429 Polycarbonate, Stainless Steel (316) i-DT61PW AM
01919430 Polycarbonate, Stainless Steel (316) i-DT61PW AM Front Installation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01919431 Supplier OHAUS™ Supplier No. 30575584

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Defender 6000 Series Extreme Washdown Indicator is specifically designed for food, chemical and pharmaceutical applications including production, processing, and packaging.

  • Applications: Weighing, Counting with APW optimization, Percent Weighing, Checkweighing, Checkcounting, Dynamic Weighing/Display Hold
  • Display: 6-digit, 7-segment LCD display with white backlight, 45 mm high digits
  • Operation: 6 D (LR20) Batteries (Included)
  • Communication: Easy access IR communication kit (available as an accessory); GLP/GMP data output with real-time clock
  • Construction: IP68/IP69K food grade polycarbonate housing and 316 stainless steel mounting bracket
  • Design Features: 3-color check light bar, menu and key lock, selectable environmental and auto-print settings, stability indicator, overload/underload indicators, low battery indicator, auto shut-off, auto tare

Specifications

Battery Life Depends on the capacity of power bank used (Up to 55 hours with 20,000 mAh power bank)
Construction Stainless Steel (316)
Display Type Backlit LCD
Model i-DT61XWE AM
Protection Class IP68 immersion/IP69K washdown
Type Bench Scales
Legal for Trade Class III/IIIL/IIIHD