Learn More
Research Products Corporation Deep Well Plates, Sterile, 96 Square V Bottom Wells, 2.2ml, 5 per Package
Numéro de catalogue. 501537648
Encompass
Deep Well Plates are ideal for sample collection, preparation and long-term storage. Raised rim on each well facilitates secure sealing with film or sealing mat. Plates allow optimum sample mixing and recovery through highly polished walls and offer reliable automation performance. Wells feature alphanumeric indexing molded in on the A-H and 1-12 axis with maximum centrifugation to 4,000 RCF.
Manufactured from virgin polypropylene (autoclavable) and are ultra-clear for easy visualization of samples. Plates are compatible with most liquid handling robots and SBS footprint meets ANSI/SLAS 1-2004. Stackable and flatness tested for reliable automation performance.
Numéro de catalogue. 501537648 Fournisseur Research Products Corporation Code fournisseur 162523S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Deep Well Plates are ideal for sample collection, preparation and long-term storage. Raised rim on each well facilitates secure sealing with film or sealing mat. Plates allow optimum sample mixing and recovery through highly polished walls and offer reliable automation performance. Wells feature alphanumeric indexing molded in on the A-H and 1-12 axis with maximum centrifugation to 4,000 RCF.
Manufactured from virgin polypropylene (autoclavable) and are ultra-clear for easy visualization of samples. Plates are compatible with most liquid handling robots and SBS footprint meets ANSI/SLAS 1-2004. Stackable and flatness tested for reliable automation performance.