Research Products Corporation Sealing Mats for Deep Well Plates, Sterile, fits 96 Square Wells, 10 per Package
Numéro de catalogue. 501537652
Sealing Mats are manufactured of durable, pierceable silicone. Designed to fit Deep Well Plates and preserve contents and are suitable for heat sealing. Sealing Mats can be rewashed and reused, but autoclaving is not recommended.
Numéro de catalogue.
501537652
Fournisseur
Research Products CorporationCode fournisseur
162525S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Sealing Mats are manufactured of durable, pierceable silicone. Designed to fit Deep Well Plates and preserve contents and are suitable for heat sealing. Sealing Mats can be rewashed and reused, but autoclaving is not recommended.