Celltreat Scientific Products 96 Deep Well Storage Plate, 1.0mL, PP, Round Well, V-Bottom, Sterile
Numéro de catalogue. NC2131779
96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications. Round Well with V-Bottom is ideal for vortexing and sample retrieval.
Numéro de catalogue.
NC2131779
Fournisseur
Celltreat Scientific ProductsCode fournisseur
229570S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications. Round Well with V-Bottom is ideal for vortexing and sample retrieval.