Learn More
GA International Deep-Freeze Cover-Up Labels for Thermal-Transfer Barcode Printers 2.5"" x 1"" / 63.5mm x 25.4mm Rectangle White From -80C to +120C (-112F to +248F)
Numéro de catalogue. 5000571411
Encompass
Deep-freeze permanent blackout labels are designed to conceal any already existing information or labels. Ideal for re-labeling cryo vials, microtubes, and freezer boxes, to be stored in ultra-low temperature freezers (-80C) or to be transported in dry ice.These cover-up labels will completely obscure the existing label and block-out the information underneath. Printable with all major brands of thermal-transfer printers, these versatile labels can withstand a wide temperature range, also suitable for high-temperature applications, as high as +120C.
Numéro de catalogue. 5000571411 Fournisseur GA International Code fournisseur BOC5C32WH
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Deep-freeze permanent blackout labels are designed to conceal any already existing information or labels. Ideal for re-labeling cryo vials, microtubes, and freezer boxes, to be stored in ultra-low temperature freezers (-80C) or to be transported in dry ice.These cover-up labels will completely obscure the existing label and block-out the information underneath. Printable with all major brands of thermal-transfer printers, these versatile labels can withstand a wide temperature range, also suitable for high-temperature applications, as high as +120C.