missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decyl Methyl Sulfide 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 mL |
| Formula Weight | 188.37 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Decyl Methyl Sulfide |
Safety and Handling
| TSCA | No |