missing translation for 'onlineSavingsMsg'
Learn More

Decyl aldehyde, 95%

Catalog No. AC154975000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154975000 500 mL Glass bottle
AC154971000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154975000 Supplier Thermo Scientific Chemicals Supplier No. 154975000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-31-2 | C10H20O | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-31-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00007031
InChI Key KSMVZQYAVGTKIV-UHFFFAOYSA-N
Synonym decyl aldehyde, capraldehyde, caprinaldehyde, decanaldehyde, n-decyl aldehyde, capric aldehyde, 1-decanal, n-decanal, n-decaldehyde, 1-decyl aldehyde
PubChem CID 8175
ChEBI CHEBI:31457
IUPAC Name decanal
SMILES CCCCCCCCCC=O

Specifications

Melting Point 7.0°C
Color Colorless to Yellow
Density 0.8300g/mL
Boiling Point 207.0°C to 209.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4270 to 1.4300
Linear Formula CH3(CH2)8CHO
Quantity 500 mL
Beilstein 01,711
Specific Gravity 0.83
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in 80% alcohol,insoluble in glycerol
Formula Weight 156.27
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Decyl aldehyde, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-957-4
RTECSNumber HD6000000
TSCA TSCA

RUO – Research Use Only