missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decon™ LeStreak™ Microstreakers
Perfect for inoculating or transferring microbiological samples
Supplier: Decon™ MP1875
Description
Perfect for inoculating or transferring microbiological samples
- Use to inoculate or transfer microbiological samples
- Made of high-quality twisted nichrome wire permanently attached to lightweight insulated aluminum handles
- Slide smoothly across agar surfaces without tearing or gouging
- Nichrome wire cools rapidly after flaming
- 1 and 10μL loops are individually calibrated in accordance with ASM/APHA via Evans Blue Dye Test
- Noncalibrated microstreakers are color coded for easy identification
Certifications
Calibration certificates provided for 1μL and 10μL loops
Specifications
| 10.75 in. | |
| 0.05 in. | |
| Nichrome | |
| 10 μL | |
| Inoculating Streaker | |
| Yes |
| Non-sterile | |
| 0.14 cm | |
| 27.31 cm | |
| Large | |
| Nichrome wire loop permanently attached to silver insulated aluminum holder, calibration certificate in accordance with ASM/APHA via evans blue test |