missing translation for 'onlineSavingsMsg'
Learn More

Decanoyl chloride, 98+%

Catalog No. AC161191000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161191000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC161191000 Supplier Thermo Scientific Chemicals Supplier No. 161191000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-13-0 | C10H19ClO | 190.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-13-0
Molecular Formula C10H19ClO
Molecular Weight (g/mol) 190.71
MDL Number MFCD00000771
InChI Key IPIVAXLHTVNRBS-UHFFFAOYSA-N
Synonym capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n
PubChem CID 66982
IUPAC Name decanoyl chloride
SMILES CCCCCCCCCC(=O)Cl

Specifications

Melting Point -34.5°C
Color Brown to White
Density 0.9190g/mL
Boiling Point 232.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420
Linear Formula CH3(CH2)8COCl
Quantity 100 g
Beilstein 02, 356
Specific Gravity 0.919
Solubility Information Solubility in water: 100mg/l (20°C). Other solubilities: hydrolyses in water,450g/L methanol- 500g/L ethanol,500g/L 2-propanol- 500g/L dmf,500g/L acetone- 500g/L dichloromethane
Formula Weight 190.71
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material Decanoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 203-938-0
TSCA TSCA

RUO – Research Use Only