missing translation for 'onlineSavingsMsg'
Learn More

Decane 99.5+%, TCI America™

Catalog No. S02825ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02825ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02825ML Supplier TCI America Supplier No. S02825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00008954
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Color Colorless
Boiling Point 174°C
Quantity 5 mL
UN Number 2247
Formula Weight 142.29
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Decane [Standard Material for GC]
EINECSNumber (2)-0010
RTECSNumber HD6550000
TSCA Yes