missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decane, 99+%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Color | Colorless |
| Density | 0.7350g/mL |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99+% |
| Packaging | Glass bottle |
| Refractive Index | 1.4103 to 1.4123 (20°C, 589nm) |
| Linear Formula | CH3(CH2)8CH3 |
| Quantity | 2.5 L |
| Beilstein | 01, 168 |
| Formula Weight | 142.28 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: May be fatal if swallowed and enters airways. Flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 204-686-4 |
| RTECSNumber | HD6550000 |
| TSCA | TSCA |
RUO – Research Use Only