missing translation for 'onlineSavingsMsg'
Learn More

Decahydro-2-naphthyl Formate (mixture of isomers) 90.0+%, TCI America™

Catalog No. D339925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D339925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D339925G Supplier TCI America Supplier No. D339925G

Chemical Identifiers

CAS 10519-12-7
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.263
MDL Number MFCD00046494
InChI Key NMFWOBQDDWFAAB-UHFFFAOYSA-N
Synonym Formic Acid Decahydro-2-naphthyl Ester
PubChem CID 25325
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalen-2-yl formate
SMILES C1CCC2CC(CCC2C1)OC=O

Specifications

Color White-Yellow
Boiling Point 126°C
Quantity 25 g
Formula Weight 182.26
Percent Purity ≥90.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Decahydro-2-naphthyl Formate (mixture of isomers)
EINECSNumber (4)-0368
RTECSNumber QL3448000
TSCA Yes