missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 1.0 N Aqueous Solution, Spectrum™ Chemical

Catalog No. 18613945
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613945 4 L Amber Glass Bottle
18613943 500 mL Amber Glass Bottle
18613944 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613945 Supplier Spectrum Chemical Mfg Cor Supplier No. S5454LTPL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 540-72-7 , 7732-18-5 , 10102-17-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
IUPAC Name sodium cyanosulfanide
SMILES [Na+].[S-]C#N

Specifications

Physical Form Solution
Quantity 4 L
Grade Ungraded
Concentration or Composition (by Analyte or Components) 1.0 N ±0.03 N
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Thiosulfate, 1.0 N Aqueous Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.