Learn More
Electron Microscopy Sciences Double Sides Carbon Conductive Tape, 25 mm x 5m 5 MT
Numéro de catalogue. 50366500
Encompass
EMS introduces this unique double sided carbon tape which is 260 microns in thickness. The base which is Polycarbonate and 200 microns thick has on both of its sides 30 microns of conductive glue giving it a total thickness of 260 microns. The tape has one transparent liner on one side and a white thicker one on the reverse which protects the tape in shipping. These are considered to be small impurities but they may be present
Width: 25 mm
Numéro de catalogue. 50366500 Fournisseur Electron Microscopy Sciences Code fournisseur 7781925
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
EMS introduces this unique double sided carbon tape which is 260 microns in thickness. The base which is Polycarbonate and 200 microns thick has on both of its sides 30 microns of conductive glue giving it a total thickness of 260 microns. The tape has one transparent liner on one side and a white thicker one on the reverse which protects the tape in shipping. These are considered to be small impurities but they may be present
Width: 25 mm