Learn More
Premier Lab Supply 32mm XRF Sample Cups PN# SC-4331
Catalog No. NC0552256
Encompass
32mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# SC-4331) Suitable for Universal OD: 32mm; ID: 24mm; Height: 26mm; Volume: 12mL 100 per bag
Catalog No. NC0552256 Supplier Premier Lab Supply Supplier No. SC4331
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
32mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# SC-4331) Suitable for Universal OD: 32mm; ID: 24mm; Height: 26mm; Volume: 12mL 100 per bag