Weber DB Sterilized Pre-filled Dilution Bottles. Butterfield's Buffer. Case of 72. Tamper evident sterility strip is integrated as part of the cap. PET bottle has glass-like clarity. White cap. 99mL fill.
Numéro de catalogue.
NC0362669
Fournisseur
Weber ScientificCode fournisseur
312714
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Weber DB Sterilized Pre-filled Dilution Bottles. Butterfield's Buffer. Case of 72. Tamper evident sterility strip is integrated as part of the cap. PET bottle has glass-like clarity. White cap. 99mL fill.