missing translation for 'onlineSavingsMsg'
Learn More

Interscience DataLink

The DataLink system transmits, prints and labels the edge of Petri dishes for easy identification.

Supplier:  Interscience 439050

 Afficher plus de versions de ce produit

Numéro de catalogue. 16948922

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

DataLink makes it easy to identify Petri dishes during its microbiological analysis. The DataLink software controls and transmits the data on a label. All the information relating to the analysis of the sample is traced and automated.

  • User account management
  • Monitoring of sample data and automation of settings
  • Audit trail, 21 CFR part 11

DataLink:

  • Label size: 50 x 10 mm

DataLink Pro:

  • Label size: 50 x 5 mm
  • Semi-automatic labelling platform
Spécifications

Identifiants chimiques

Labeling Petri Dishes
DataLink

Spécifications

Labeling Petri Dishes
DataLink
DataLink Pro
100/240 V
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions