missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Daptomycin

Catalog No. 391710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
391710 10 mg
391750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 391710 Supplier Tocris Bioscience™ Supplier No. 3917/10

Antibiotic; inhibits protein, DNA and RNA synthesis in gram-positive bacteria

Daptomycin is a lipopeptide, calcium-dependent antibiotic. Exhibits potent bacteriocidal activity against most gram-positive bacteria in vitro and in vivo, including antibiotic-resistant strains such as MRSA and VRE. Disrupts plasma membrane function; activity results in membrane depolarization leading to inhibition of protein, DNA and RNA synthesis.

Chemical Identifiers

CAS 103060-53-3
Molecular Formula C72H101N17O26

Specifications

Quantity 10 mg
Formula Weight 1620.67
Product Type Daptomycin
Purity >95%
Molecular Weight (g/mol) 1620.693
Chemical Name or Material Daptomycin
Recommended Storage Store at -20°C