accessibility menu, dialog, popup

UserName

H3COHN(S)NHONH(S)NH2OOHN(S)OHOONHH3COO(S)ONH2NHO(S)HNO(R)OHNHOHNO(S)HOOHNO(R)CH3HNO(S)OHONHO(S)H2NHNONHOH3CHOO

CAS RN 103060-53-3

IUPAC Name: (3S)-3-{[(3S,6S,9R,15S,18R,21S,24S)-3-[2-(2-aminophenyl)-2-oxoethyl]-24-(3-aminopropyl)-15,21-bis(carboxymethyl)-6-(1-carboxypropan-2-yl)-9-(hydroxymethyl)-18,31-dimethyl-2,5,8,11,14,17,20,23,26,29-decaoxo-1-oxa-4,7,10,13,16,19,22,25,28-nonaazacyclohentriacontan-30-yl]carbamoyl}-3-[(2S)-3-carbamoyl-2-[(2S)-2-decanamido-3-(1H-indol-3-yl)propanamido]propanamido]propanoic acid
Synonyms: daptomycin
Molecular Weight (g/mol): 1620.69
Molecular Formula: C72H101N17O26
InChi Key: DOAKLVKFURWEDJ-OFNKPWESSA-N
SMILES: CCCCCCCCCC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)NC1C(C)OC(=O)[C@H](CC(=O)C2=CC=CC=C2N)NC(=O)[C@@H](NC(=O)[C@@H](CO)NC(=O)CNC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCN)NC(=O)CNC1=O)C(C)CC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Product Type 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)