missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daptomycin 94.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Quantity | 100 mg |
| Formula Weight | 1620.69 |
| Percent Purity | ≥94.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Daptomycin |
Sécurité et manipulation
| RTECSNumber | HB5626000 |
| TSCA | No |
| Recommended Storage | Freezer (-20 °C) |