missing translation for 'onlineSavingsMsg'
Learn More

DAPT 97.0+%, TCI America™

Catalog No. D425725MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D425725MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D425725MG Supplier TCI America Supplier No. D425725MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 208255-80-5
Molecular Formula C23H26F2N2O4
Molecular Weight (g/mol) 432.468
MDL Number MFCD04974585
InChI Key DWJXYEABWRJFSP-XOBRGWDASA-N
Synonym dapt, dapt gsi-ix, gamma-secretase inhibitor ix, gsi-ix, insolution gamma-secretase inhibitor ix, s-tert-butyl 2-s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, n-n-3,5-difluorophenacetyl-l-alanyl-s-phenylglycine t-butyl ester, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetyl amino propanoyl amino-2-phenylacetate, gsiix
PubChem CID 5311272
ChEBI CHEBI:86193
IUPAC Name tert-butyl (2S)-2-[[(2S)-2-[[2-(3,5-difluorophenyl)acetyl]amino]propanoyl]amino]-2-phenylacetate
SMILES CC(C(=O)NC(C1=CC=CC=C1)C(=O)OC(C)(C)C)NC(=O)CC2=CC(=CC(=C2)F)F

Specifications

Color White-Yellow
Quantity 25 mg
Formula Weight 432.47
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material DAPT
TSCA No
Recommended Storage Refrigerator