missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ DAPT
Description
DAPT is an ancillary Material Grade version of DAPT (Cat. No. 2634). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.
Find out more about our Ancillary Material Grade product range.
Specifications
Specifications
| Chemical Name or Material | (2 S)-N-[(3,5-Difluorophenyl)acetyl]-L-alanyl-2-phenylglycine 1,1-dimethylethyl ester |
| CAS | 208255-80-5 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Secretase Inhibitors |
| Molecular Formula | C23H26F2N2O4 |
| Purity | 0.99 |