missing translation for 'onlineSavingsMsg'
Learn More

Dansylglycine 98.0+%, TCI America™

Catalog No. D15015G
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15015G 5 g
D1501100MG 100 mg
D15011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D15015G Supplier TCI America Supplier No. D15015G

Chemical Identifiers

CAS 1091-85-6
Molecular Formula C14H16N2O4S
Molecular Weight (g/mol) 308.352
MDL Number MFCD00037734
InChI Key QHFXORCWAQTTGH-UHFFFAOYSA-N
Synonym N-[5-(Dimethylamino)naphthalene-1-sulfonyl]glycine, Dns-Gly-OH
PubChem CID 70666
IUPAC Name 2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]acetic acid
SMILES CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)NCC(=O)O

Specifications

Melting Point 159°C
Quantity 5 g
Formula Weight 308.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dansylglycine