missing translation for 'onlineSavingsMsg'
Learn More

Dansylamino-PITC 98.0+%, TCI America™

Catalog No. A5354100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5354100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5354100MG Supplier TCI America Supplier No. A5354100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102417-94-7
Molecular Formula C19H17N3O2S2
Molecular Weight (g/mol) 383.48
MDL Number MFCD00077739
InChI Key MZGXHCHKGRQLHR-UHFFFAOYSA-N
Synonym 4-(Dansylamino)phenyl Isothiocyanate, 4-[[5-(Dimethylamino)-1-naphthylsulfonyl]amino]phenyl Isothiocyanate, DNSAPITC
PubChem CID 128100
IUPAC Name 5-(dimethylamino)-N-(4-isothiocyanatophenyl)naphthalene-1-sulfonamide
SMILES CN(C)C1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 129°C
Quantity 100 mg
Formula Weight 383.48
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Dansylamino-PITC [Fluorescent Coupling Reagent for Edman Degradation]
TSCA No
Recommended Storage Refrigerator