missing translation for 'onlineSavingsMsg'
Learn More

Dansyl Chloride 98.0+%, TCI America™

Catalog No. D06565G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06565G 5 g
D06561G 1 g
D065625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D06565G Supplier TCI America Supplier No. D06565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 605-65-2
Molecular Formula C12H12ClNO2S
Molecular Weight (g/mol) 269.74
MDL Number MFCD00003985
InChI Key XPDXVDYUQZHFPV-UHFFFAOYSA-N
Synonym dansyl chloride, 5-dimethylamino naphthalene-1-sulfonyl chloride, dns chloride, dansyl, 5-dimethylamino-1-naphthalenesulfonyl chloride, 1-naphthalenesulfonyl chloride, 5-dimethylamino, 1-dimethylaminonaphthalene-5-sulfonyl chloride, 1-chlorosulfonyl-5-dimethylaminonaphthalene, unii-qmu9166tj4, 5-dimethylaminonaphthalene-1-sulphonyl chloride
PubChem CID 11801
ChEBI CHEBI:51907
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonyl chloride
SMILES CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O

Specifications

Melting Point 70°C
Quantity 5 g
UN Number 3261
Formula Weight 269.74
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dansyl Chloride
RTECSNumber QK3688000
TSCA Yes
Recommended Storage Refrigerator