missing translation for 'onlineSavingsMsg'
Learn More

Dansyl Chloride (10% in Acetone), TCI America™

Catalog No. D000510ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D000510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D000510ML Supplier TCI America Supplier No. D000510ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 605-65-2
Molecular Formula C12H12ClNO2S
Molecular Weight (g/mol) 269.74
MDL Number MFCD00003985
InChI Key XPDXVDYUQZHFPV-UHFFFAOYSA-N
Synonym 5-(Dimethylamino)naphthalene-1-sulfonyl Chloride
PubChem CID 11801
ChEBI CHEBI:51907
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonyl chloride
SMILES CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O

Specifications

UN Number 2924
Quantity 10 mL
Physical Form Liquid
Formula Weight 269.74
Chemical Name or Material Dansyl Chloride (10% in Acetone)
RTECSNumber QK3688000
TSCA Yes
Recommended Storage Refrigerator