missing translation for 'onlineSavingsMsg'
Learn More

Daidzein 98.0+%, TCI America™

Catalog No. D26681G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26681G Supplier TCI America Supplier No. D26681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 486-66-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00016954
InChI Key ZQSIJRDFPHDXIC-UHFFFAOYSA-N
Synonym daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
PubChem CID 5281708
ChEBI CHEBI:28197
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 254.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Daidzein
RTECSNumber DJ3100040
TSCA No