missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ DAF-FM DA, Calbiochem™,

Catalog No. 2515201MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2515201MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2515201MG Supplier MilliporeSigma™ Supplier No. 2515201MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cell-permeable, photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of approx. 3nM

Chemical Identifiers

CAS 254109-22-3
Molecular Formula C25H18F2N2O7
Molecular Weight (g/mol) 496.42
MDL Number MFCD16872050
InChI Key BEVHTVRRVVEMEF-UHFFFAOYSA-N
Synonym daf-fm da, 3-amino-4-n-methylamino-2', 7'-difluorofluorescein diacetate, 3-amino, 4-aminomethyl-2′,7′-difluorofluorescein diacetate, daf-fm da cell permeable, daf-fm da cellpermeable, diaminofluorescein-fm diacetate, daf-fm da hplc, 4-amino-5-methylamino-2',7'-difluorescein diacetate, 4-amino-5-methylamino-2',7'-difluorofluorescein diacetate, daf-fm da solution 5 mm in dmso , 1 mg in 0.40 ml dmso
PubChem CID 2762646
IUPAC Name 6'-(acetyloxy)-4-amino-2',7'-difluoro-5-(methylamino)-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-3'-yl acetate
SMILES CNC1=C(N)C2=C(C=C1)C1(OC2=O)C2=CC(F)=C(OC(C)=O)C=C2OC2=CC(OC(C)=O)=C(F)C=C12

Specifications

Color Yellow
Quantity 1 mg
Formula Weight 496.4
Percent Purity ≥98%
Grade HPLC
Concentration or Composition (by Analyte or Components) 5mM solution in DMSO
Physical Form Liquid
Chemical Name or Material DAF-FM DA
RTECSNumber PV6210000
Recommended Storage -20°C (-4°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).