missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ DAF-FM, Calbiochem™,

Catalog No. 2515151MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2515151MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2515151MG Supplier MilliporeSigma™ Supplier No. 2515151MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of approx. 3nM

Chemical Identifiers

CAS 254109-20-1
Molecular Formula C21H14F2N2O5
Molecular Weight (g/mol) 412.349
InChI Key DIJCILWNOLHJCG-UHFFFAOYSA-N
Synonym daf-fm, 3-amino, 4-aminomethyl-2′,7′-difluorofluorescein, daf-fm hplc, 4-amino-5-methylamino-2',7'-difluorescein, 4-amino-5-methylamino-2',7'-difluorofluorescein, 7-amino-2',7'-difluoro-3',6'-dihydroxy-6-methylamino spiro, 4-amino-2',7'-difluoro-3',6'-dihydroxy-5-methylamino spiro 2-benzofuran-1,9'-xanthen-3-one, 4-amino-5-n-methylamino-2',7'-difluoro-3',6'-dihydroxy-spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 4-amino-5-methylamino-2 inverted exclamation marka,7 inverted exclamation marka-difluorescein
PubChem CID 10431792
IUPAC Name 7-amino-2',7'-difluoro-3',6'-dihydroxy-6-(methylamino)spiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES CNC1=C(C2=C(C=C1)C3(C4=CC(=C(C=C4OC5=CC(=C(C=C53)F)O)O)F)OC2=O)N

Specifications

Color Yellow
Quantity 1 mg
Formula Weight 412.4
Percent Purity ≥98%
Grade HPLC
Concentration or Composition (by Analyte or Components) 5mM solution in DMSO
Physical Form Liquid
Chemical Name or Material DAF-FM
RTECSNumber PV6210000
Recommended Storage -20°C (-4°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).