MW 373.32 g/mol, Purity >95%. Highly fluorescent reference material for DAF-2 (ab145269). DAF-2 is membrane impermeable and reacts with Nitric Oxide to generate the highly fluorescent triazole DAF-2T.
The product is subject to the following: Abcam Restricted Use Statement