missing translation for 'onlineSavingsMsg'
Learn More

Electron Microscopy Sciences (DAB), 3,3'-Diaminobenzidine Purified free-base, EM Grade 1 GR

Numéro de catalogue. 50980347
Encompass

(3,3',4,4'-Biphenyltetramin)
CAS #91-95-2 CHN F.W. 214.27
Melting point: 173C

Used as an ultrastructural protein tracer for immuno-cytochemistry. A possible carcinogen. Purified free-base.
Grade: EM Grade

Numéro de catalogue. 50980347 Fournisseur Electron Microscopy Sciences Code fournisseur 13060

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

(3,3',4,4'-Biphenyltetramin)
CAS #91-95-2 CHN F.W. 214.27
Melting point: 173C

Used as an ultrastructural protein tracer for immuno-cytochemistry. A possible carcinogen. Purified free-base.
Grade: EM Grade