missing translation for 'onlineSavingsMsg'
Learn More

D-Valinol 98.0+%, TCI America™

Numéro de catalogue. p-7137915
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
V00771G 1 g
V00775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. V00771G Fournisseur TCI America Code fournisseur V00771G
Il en reste null

Identifiants chimiques

CAS 4276-09-9
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00064297
InChI Key NWYYWIJOWOLJNR-YFKPBYRVSA-N
Synonym d-valinol, r---2-amino-3-methyl-1-butanol, r-2-amino-3-methylbutan-1-ol, 2r-2-amino-3-methylbutan-1-ol, r-2-amino-3-methyl-1-butanol, d-2-amino-3-methyl-1-butanol, 2r-2-amino-3-methyl-1-butanol, 1r-1-isopropyl-2-hydroxyethylamine, d---valinol r---2-amino-3-methyl-1-butanol, d-valinol r---2-amino-3-methyl-1-butanol
PubChem CID 6950587
IUPAC Name (2R)-2-amino-3-methylbutan-1-ol
SMILES CC(C)[C@@H](N)CO

Spécifications

Color White-Yellow
Quantity 1 g
Formula Weight 103.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material D-Valinol