missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Valine, 98+%

Catalog No. AAA1889422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1889422 100 g
AAA1889406 5 g
AAA1889414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1889422 Supplier Thermo Scientific Chemicals Supplier No. A1889422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 640-68-6 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 640-68-6
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064219
InChI Key KZSNJWFQEVHDMF-SCSAIBSYSA-N
Synonym d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid
PubChem CID 71563
ChEBI CHEBI:27477
IUPAC Name (2R)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@@H](N)C(O)=O

Specifications

Melting Point 295°C (decomposition)
Beilstein 1721135
Quantity 100 g
Formula Weight 117.15
Optical Rotation −27° (c=5 in 5N HCl)
Percent Purity ≥98%
Chemical Name or Material D-Valine
EINECSNumber 211-368-9
RTECSNumber YV9360000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only