missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Valine, 98+%

Catalog No. AC227220250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227220250 25 g Glass Bottle
AC227220050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC227220250 Supplier Thermo Scientific Chemicals Supplier No. 227220250

CAS: 640-68-6 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 640-68-6
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064219
InChI Key KZSNJWFQEVHDMF-SCSAIBSYSA-N
Synonym d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid
PubChem CID 71563
ChEBI CHEBI:27477
IUPAC Name (2R)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@@H](N)C(O)=O

Specifications

Melting Point 302.0°C to 303.0°C
Color White
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (105°C, 4 hrs)
Assay Percent Range 98+%
Packaging Glass Bottle
Linear Formula (CH3)2CHCH(NH2)CO2H
Specific Rotation − 27.50
Sulfated Ash 0.2% max.
Solubility Information Solubility in water: 56g/L soluble in water (20°C). Other solubilities: soluble in mineral acids, insoluble in organic solvents, soluble in ethanol, diethyl ether, benzene
Water 0.5% max.
Specific Rotation Condition −27.50 (20.00°C c=5,5N HCl)
Quantity 25 g
Formula Weight 117.15
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material D-Valine, 98+%
Show More Show Less
EINECSNumber 211-368-9
RTECSNumber YV9360000
TSCA TSCA

RUO – Research Use Only