missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Tyrosine tert-butyl ester, 98%
Description
D-Tyrosine tert-butyl ester, is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Tyrosine tert-butyl ester, is used as a pharmaceutical intermediate.
Solubility
Very slightly soluble in cold water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
D-Tyrosine tert-butyl ester, is used as a pharmaceutical intermediate.
Solubility
Very slightly soluble in cold water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Specifications
| Solubility Information | Very slightly soluble in cold water. |
| Quantity | 5 g |
| Formula Weight | 237.14 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | D-Tyrosine tert-butyl ester |
Safety and Handling
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only