missing translation for 'onlineSavingsMsg'
Learn More

D-Tyrosine, 99%

Catalog No. AAA1770914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1770914 25 g
AAA1770903 1 g
AAA1770906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1770914 Supplier Thermo Scientific Chemicals Supplier No. A1770914

CAS: 556-02-5 | C9H11NO3 | 181.19 g/mol

D-Tyrosine is synthesized from phenylalanine. It is also the precursor of epinephrine, thyroid hormones, and melanin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tyrosine is synthesized from phenylalanine. It is also the precursor of epinephrine, thyroid hormones, and melanin.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 556-02-5
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00063073
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine
PubChem CID 71098
ChEBI CHEBI:28479
IUPAC Name (2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point 310°C to 314°C
Beilstein 2212157
Merck Index 14,9839
Solubility Information Soluble in water.
Quantity 25 g
Formula Weight 181.19
Optical Rotation +11° (c=4 in 1N HCl)
Percent Purity 99%
Chemical Name or Material D-Tyrosine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-112-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only