missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Trehalose, Dihydrate, Spectrum™ Chemical

Catalog No. 18602004
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
50 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602004 50 kg Amber Glass Bottle
18601998 5 g Amber Glass Bottle
18601999 25 g Amber Glass Bottle
18602000 100 g Amber Glass Bottle
18602001 500 g Amber Glass Bottle
18602002 1 kg Amber Glass Bottle
18602003 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18602004 Supplier Spectrum Chemical Mfg Cor Supplier No. T109750KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

T1097, 6138-23-4, C12H22O11·2H2O

Spectrum™ Chemical D-(+)-Trehalose, Dihydrate , is a natural alpha-linked disaccharide that is also referred to as mycose or tremalose. It is found naturally in plants and microorganisms such as grasshoppers, shrimp, butterflies and bees. It is thought to have a use in the treatment for Huntington's and Parkinson's disease as it possibly corrects defects in autophagy seen in these diseases.

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.33
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-DJCWUSJTNA-N
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
SMILES O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Packaging Amber Glass Bottle
Quantity 50 kg
Specific Rotation +197° to +201°
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material D-(+)-Trehalose, Dihydrate