missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Threonine 98.0+%, TCI America™

Catalog No. T0228500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0228500G 500 g
T022825G 25 g
T0228100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0228500G Supplier TCI America Supplier No. T0228500G
Only null left

Chemical Identifiers

CAS 632-20-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064269
InChI Key AYFVYJQAPQTCCC-STHAYSLISA-N
Synonym d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth
PubChem CID 69435
ChEBI CHEBI:16398
IUPAC Name (2R,3S)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](N)C(O)=O

Specifications

Color White
Quantity 500 g
Formula Weight 119.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Threonine
EINECSNumber (9)-1584
RTECSNumber XO8580000
TSCA No