missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Tartaric Acid 99.0+%, TCI America™

Catalog No. T0026500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0026500G 500 g
T002625G 25 g
T0026100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0026500G Supplier TCI America Supplier No. T0026500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 147-71-7
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00004238
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, #NAME?, 2s,3s-2,3-dihydroxybutanedioic acid, #NAME?, s,s-tartaric acid, 2s,3s---tartaric acid
PubChem CID 439655
ChEBI CHEBI:15672
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Color White
Quantity 500 g
Physical Form Crystalline Powder
Formula Weight 150.09
Percent Purity ≥99.0% (T)
Chemical Name or Material D-(-)-Tartaric Acid
EINECSNumber (2)-1456
RTECSNumber WW7875000
TSCA Yes