missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Tartaric acid, 99%

Catalog No. AAA1126436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1126436 500 g
AAA1126414 25 g
AAA1126422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1126436 Supplier Thermo Scientific Chemicals Supplier No. A1126436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 147-71-7 | C4H6O6 | 150.09 g/mol

D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.

Solubility
Soluble in water and alcohol.

Notes
Light sensitive. Incompatible with bases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 147-71-7
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00004238
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acid
PubChem CID 439655
ChEBI CHEBI:15672
IUPAC Name (2S,3S)-2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

pH 1.6
Melting Point 166°C to 170°C
Quantity 500 g
Flash Point 210°C (410°F)
Odor Odorless
Optical Rotation −13° (c=10 in water)
Beilstein 1725145
Merck Index 14,9068
Solubility Information Soluble in water and alcohol.
Formula Weight 150.09
Percent Purity 99%
Chemical Name or Material D- (-)-Tartaric acid
Show More Show Less
Hazard Category H318-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 205-695-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only