missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Talose 98.0+%, TCI America™

Catalog No. T0869500MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0869500MG 500 mg
T0869100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0869500MG Supplier TCI America Supplier No. T0869500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2595-98-4
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00135834
InChI Key GZCGUPFRVQAUEE-UHFFFAOYNA-N
Synonym d-+-talose, aldehydo-d-talose, 2s,3s,4s,5r-2,3,4,5,6-pentahydroxyhexanal
PubChem CID 99459
ChEBI CHEBI:68461
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal
SMILES OCC(O)C(O)C(O)C(O)C=O

Specifications

Melting Point 134°C
Color White
Quantity 500 mg
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Talose