missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Sorbitol, Anhydrous, Cell Culture Reagent, ≥98%, MP Biomedicals™
D-Sorbitol, Anhydrous, Cell Culture Reagent, ≥98% is used for inducing osmotic stress.
$112.43
Chemical Identifiers
| CAS | 50-70-4 |
|---|---|
| Molecular Formula | C6H14O6 |
| Molecular Weight (g/mol) | 182.17 |
| InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
| Synonym | d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, glucarine, diakarmon, multitol, sorbilande |
| PubChem CID | 5780 |
| ChEBI | CHEBI:17924 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN19474201
|
N/A | 1 kg |
Each for $112.43
|
|
|||||
Description
- Used for washing spheroplasts and in isoelectric focusing to minimize endoosmotic flow in agarose gels.
- May be used to induce osmotic stress.
Chemical Identifiers
| 50-70-4 | |
| 182.17 | |
| d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, glucarine, diakarmon, multitol, sorbilande | |
| CHEBI:17924 |
| C6H14O6 | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| 5780 |
Specifications
| 50-70-4 | |
| 1.49 g/cm3 at 59°F | |
| Very High | |
| C6H14O6 | |
| Water Solubility: 2,750,000 mg/L at 30°C | |
| −4 to 0 ° (c=2, water) | |
| 5780 | |
| 9.9X10-9 mm Hg at 25°C | |
| Cell Culture Grade | |
| <20.0 μS/cm |
| 230°F | |
| 7.0 | |
| 542°F | |
| d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, glucarine, diakarmon, multitol, sorbilande | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| 182.17 | |
| CHEBI:17924 | |
| ≥98% | |
| Powder |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.