missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Serine, 99%

Catalog No. AAA1135306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1135306 5 g
AAA1135314 25 g
AAA1135322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1135306 Supplier Thermo Scientific Chemicals Supplier No. A1135306
Only null left

CAS: 312-84-5 | C3H7NO3 | 105.093 g/mol

D-Serine is involved in the biosynthesis of purines, pyrimidines, and other amino acids. D-Serine also is an agonist of glycine site of the NMDA-type glutamate receptor. It also acts as Lacosamide intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Serine is involved in the biosynthesis of purines, pyrimidines, and other amino acids. D-Serine also is an agonist of glycine site of the NMDA-type glutamate receptor. It also acts as Lacosamide intermediate.

Solubility
Soluble in water (346 mg/ml at 20°C), and mineral acids. Insoluble in alcohol, ether, and benzene.

Notes
Store at 4°C. Store away from oxidizing agents. Keep container closed when not in use.

Chemical Identifiers

CAS 312-84-5
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00004269
InChI Key MTCFGRXMJLQNBG-UWTATZPHSA-N
Synonym d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin
PubChem CID 71077
ChEBI CHEBI:16523
IUPAC Name (2R)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 218°C to 220°C (decomposition)
Assay Percent Range 99%
Beilstein 1721403
Merck Index 14,8460
Solubility Information Soluble in water (346mg/ml at 20°C),and mineral acids. Insoluble in alcohol,ether,and benzene.
Quantity 5 g
Formula Weight 105.09
Percent Purity 99%
Chemical Name or Material D-Serine
EINECSNumber 206-229-4
RTECSNumber VT8200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only