missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-Ribose 98.0+%, TCI America™
Supplier: TCI America R0025250G
| Quantity | 250 g |
|---|
Chemical Identifiers
| C5H10O5 | |
| MFCD00135453 | |
| 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917 | |
| CHEBI:47014 | |
| OC[C@H]1OC(O)[C@H](O)[C@@H]1O |
Specifications
| D-(-)-Ribose | |
| White | |
| 250 g | |
| 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917 | |
| OC[C@H]1OC(O)[C@H](O)[C@@H]1O | |
| 150.13 | |
| CHEBI:47014 | |
| ≥98.0% (GC) |
| 50-69-1 | |
| C5H10O5 | |
| MFCD00135453 | |
| HMFHBZSHGGEWLO-ZHZWXSSZNA-N | |
| (3R,4S,5R)-5-(hydroxymethyl)oxolane-2,3,4-triol | |
| 5311110 | |
| 150.13 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (8)-0048
RTECSNumber : VJ2275000
TSCA : Yes