missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Proline, 99+%

Catalog No. AC228660250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228660250 25 g Glass Bottle
AC228660010 1 g Glass Bottle
AC228660050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC228660250 Supplier Thermo Scientific Chemicals Supplier No. 228660250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 344-25-2 | C5H9NO2 | 115.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 344-25-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00064317
InChI Key ONIBWKKTOPOVIA-SCSAIBSYSA-N
Synonym d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-proline
PubChem CID 8988
ChEBI CHEBI:16313
IUPAC Name (2R)-pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O

Specifications

Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 1 % max. (105°C, 3 hrs)
Other Amino Acids 0.5% max. (TLC)
Packaging Glass Bottle
Beilstein 22, 2
Specific Rotation +83.0° to +87.0° (20°C, 589nm) (c=4, water)
Fieser 07,307; 09,393
Sulfated Ash 0.1 % max.
Specific Rotation Condition +84.75 (20.00°C c=4,H2O)
Quantity 25 g
Formula Weight 115.13
Percent Purity 99.0 to 101.0% (Total base)
Physical Form Powder
Chemical Name or Material D(+)-Proline
Show More Show Less

RUO – Research Use Only